Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256796.1 | NP_001243725.1 | 6 - 7 | 880 | 65 |
NM_001261398.1 | NP_001248327.1 | 5 - 6 | 832 | 65 | |
NM_002560.2 | NP_002551.2 | 5 - 6 | 832 | 65 | |
NR_046372.1 | 6 - 7 | 870 | 65 | ||
NR_046373.1 | 4 - 5 | 684 | 65 | ||
GenBank mRNA | AF000234.1 | - | 5 - 6 | 428 | 65 |
AK225604.1 | - | 5 - 6 | 528 | 65 | |
AK297704.1 | - | 4 - 5 | 416 | 65 | |
AK301080.1 | - | 5 - 6 | 516 | 65 | |
AK308549.1 | - | 6 - 7 | 602 | 65 | |
BC017458.1 | - | 5 - 6 | 561 | 65 | |
BC033826.1 | - | 5 - 6 | 552 | 65 | |
BT019738.1 | - | 5 - 6 | 528 | 65 | |
BX402887.2 | - | 6 - 7 | 650 | 65 | |
U83993.1 | - | 5 - 6 | 837 | 65 | |
Y07684.1 | - | 5 - 6 | 555 | 65 |