Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 63655528-63656060 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SERPINB3
Entrez Gene ID
6317
Gene Name
serpin family B member 3
Gene Aliases
HsT1196, SCC, SCCA-1, SCCA-PD, SCCA1, SSCA1, T4-A
UniGene
Hs.227948
Gene Chromosome Location
Chr.18: 63655197-63661963 on Build GRCh38
Transcripts
NM_006919.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6083438 c.857G>C p.R286P Chr.18: 63655973-63655973 on Build GRCh38
4459972 c.1149C>T p.F383F Chr.18: 63655681-63655681 on Build GRCh38
6989625 c.993G>A p.K331K Chr.18: 63655837-63655837 on Build GRCh38
6083440 c.1012G>C p.E338Q Chr.18: 63655818-63655818 on Build GRCh38
4567862 c.902_903CC>TT p.T301I Chr.18: 63655927-63655928 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs980341256 Chr.18: 63655584-63655584 on Build GRCh38
rs1055589080 Chr.18: 63655997-63655997 on Build GRCh38
rs778092071 Chr.18: 63655766-63655766 on Build GRCh38
rs759286115 Chr.18: 63656005-63656005 on Build GRCh38
rs745523433 Chr.18: 63656032-63656032 on Build GRCh38
rs777250543 Chr.18: 63655892-63655892 on Build GRCh38
rs1020966179 Chr.18: 63655874-63655874 on Build GRCh38
rs528610708 Chr.18: 63655553-63655553 on Build GRCh38
rs542903928 Chr.18: 63655860-63655860 on Build GRCh38
rs766660887 Chr.18: 63655750-63655750 on Build GRCh38
rs386804125 Chr.18: 63655778-63655780 on Build GRCh38
rs61703421 Chr.18: 63655615-63655615 on Build GRCh38
rs61748838 Chr.18: 63655994-63655994 on Build GRCh38
rs373495000 Chr.18: 63655608-63655608 on Build GRCh38
rs866537062 Chr.18: 63655863-63655863 on Build GRCh38
rs564270544 Chr.18: 63655896-63655896 on Build GRCh38
rs781029396 Chr.18: 63655975-63655975 on Build GRCh38
rs140650845 Chr.18: 63655868-63655868 on Build GRCh38
rs780920797 Chr.18: 63655778-63655778 on Build GRCh38
rs890281172 Chr.18: 63655634-63655634 on Build GRCh38
rs139301901 Chr.18: 63655993-63655993 on Build GRCh38
rs774018461 Chr.18: 63655748-63655748 on Build GRCh38
rs111442409 Chr.18: 63655788-63655788 on Build GRCh38
rs758604110 Chr.18: 63655809-63655809 on Build GRCh38
rs766656312 Chr.18: 63655927-63655927 on Build GRCh38
rs150069636 Chr.18: 63656025-63656025 on Build GRCh38
rs372901274 Chr.18: 63655904-63655904 on Build GRCh38
rs541889429 Chr.18: 63655616-63655616 on Build GRCh38
rs762603269 Chr.18: 63655654-63655654 on Build GRCh38
rs142869415 Chr.18: 63655835-63655835 on Build GRCh38
rs61733404 Chr.18: 63655739-63655739 on Build GRCh38
rs754315039 Chr.18: 63655754-63655754 on Build GRCh38
rs368675553 Chr.18: 63656006-63656006 on Build GRCh38
rs778861379 Chr.18: 63655882-63655882 on Build GRCh38
rs756523679 Chr.18: 63655661-63655661 on Build GRCh38
rs199869907 Chr.18: 63656007-63656007 on Build GRCh38
rs147015574 Chr.18: 63655915-63655915 on Build GRCh38
rs780564357 Chr.18: 63655663-63655663 on Build GRCh38
rs746777683 Chr.18: 63655712-63655712 on Build GRCh38
rs765634303 Chr.18: 63655657-63655659 on Build GRCh38
rs73962331 Chr.18: 63655764-63655764 on Build GRCh38
rs746575018 Chr.18: 63655965-63655965 on Build GRCh38
rs764001253 Chr.18: 63656016-63656016 on Build GRCh38
rs776590214 Chr.18: 63655720-63655720 on Build GRCh38
rs747762743 Chr.18: 63655838-63655838 on Build GRCh38
rs758341985 Chr.18: 63656009-63656009 on Build GRCh38
rs201128003 Chr.18: 63655914-63655914 on Build GRCh38
rs756963546 Chr.18: 63655973-63655973 on Build GRCh38
rs770652430 Chr.18: 63655715-63655715 on Build GRCh38
rs769610583 Chr.18: 63655773-63655773 on Build GRCh38
rs749645035 Chr.18: 63655980-63655980 on Build GRCh38
rs995894588 Chr.18: 63655818-63655818 on Build GRCh38
rs745614654 Chr.18: 63655823-63655823 on Build GRCh38
rs1027306367 Chr.18: 63655666-63655666 on Build GRCh38
rs1065205 Chr.18: 63655761-63655761 on Build GRCh38
rs540991572 Chr.18: 63655869-63655869 on Build GRCh38
rs60286570 Chr.18: 63655780-63655780 on Build GRCh38
rs975491056 Chr.18: 63655947-63655947 on Build GRCh38
rs369697963 Chr.18: 63655774-63655774 on Build GRCh38
rs754178743 Chr.18: 63655615-63655616 on Build GRCh38
rs755837972 Chr.18: 63655780-63655781 on Build GRCh38
rs1047257 Chr.18: 63655631-63655631 on Build GRCh38
rs753692328 Chr.18: 63655876-63655876 on Build GRCh38
rs763567283 Chr.18: 63655867-63655867 on Build GRCh38
rs769288329 Chr.18: 63655736-63655736 on Build GRCh38
rs759769723 Chr.18: 63655861-63655861 on Build GRCh38
rs780860195 Chr.18: 63655770-63655770 on Build GRCh38
rs143634391 Chr.18: 63655989-63655989 on Build GRCh38
rs778873893 Chr.18: 63655692-63655692 on Build GRCh38
rs771592861 Chr.18: 63655850-63655850 on Build GRCh38
rs775980323 Chr.18: 63655903-63655903 on Build GRCh38
rs373663669 Chr.18: 63655880-63655880 on Build GRCh38
rs754336126 Chr.18: 63655674-63655674 on Build GRCh38
rs1045467194 Chr.18: 63655840-63655840 on Build GRCh38
rs111333643 Chr.18: 63655925-63655925 on Build GRCh38
rs201374310 Chr.18: 63655780-63655780 on Build GRCh38
rs781539008 Chr.18: 63655808-63655808 on Build GRCh38
rs200813295 Chr.18: 63655866-63655866 on Build GRCh38
rs778382802 Chr.18: 63655837-63655837 on Build GRCh38
rs751744304 Chr.18: 63655795-63655795 on Build GRCh38
rs138078893 Chr.18: 63655983-63655983 on Build GRCh38
rs764272696 Chr.18: 63655794-63655794 on Build GRCh38
rs778667573 Chr.18: 63655933-63655933 on Build GRCh38
rs763789330 Chr.18: 63655659-63655659 on Build GRCh38
rs573890174 Chr.18: 63655852-63655852 on Build GRCh38
rs761063030 Chr.18: 63655749-63655749 on Build GRCh38
rs536875393 Chr.18: 63655824-63655824 on Build GRCh38
rs1001283829 Chr.18: 63655996-63655996 on Build GRCh38
rs747263941 Chr.18: 63655767-63655767 on Build GRCh38
rs190601396 Chr.18: 63655995-63655995 on Build GRCh38
rs765840103 Chr.18: 63655759-63655759 on Build GRCh38
rs971594432 Chr.18: 63655582-63655582 on Build GRCh38
rs539147734 Chr.18: 63655951-63655951 on Build GRCh38
rs376412536 Chr.18: 63655832-63655832 on Build GRCh38
rs906965433 Chr.18: 63655842-63655842 on Build GRCh38
rs763475449 Chr.18: 63655911-63655911 on Build GRCh38
rs779847786 Chr.18: 63655802-63655803 on Build GRCh38
rs748705325 Chr.18: 63655991-63655991 on Build GRCh38
rs150575224 Chr.18: 63655990-63655990 on Build GRCh38
rs755020901 Chr.18: 63655689-63655689 on Build GRCh38
rs780976447 Chr.18: 63655916-63655916 on Build GRCh38
rs750555422 Chr.18: 63655918-63655918 on Build GRCh38
rs747052084 Chr.18: 63655981-63655981 on Build GRCh38
rs141091024 Chr.18: 63655709-63655709 on Build GRCh38
rs773929167 Chr.18: 63655775-63655775 on Build GRCh38
rs752710713 Chr.18: 63655936-63655936 on Build GRCh38
rs544957272 Chr.18: 63655562-63655562 on Build GRCh38
rs753601402 Chr.18: 63655928-63655928 on Build GRCh38
rs374190073 Chr.18: 63655789-63655789 on Build GRCh38
rs751020780 Chr.18: 63655677-63655677 on Build GRCh38
rs759037224 Chr.18: 63655650-63655650 on Build GRCh38
rs759985981 Chr.18: 63655783-63655783 on Build GRCh38
rs201183606 Chr.18: 63655691-63655691 on Build GRCh38
rs771937049 Chr.18: 63655611-63655611 on Build GRCh38
rs769440445 Chr.18: 63655999-63655999 on Build GRCh38
rs559175012 Chr.18: 63655865-63655865 on Build GRCh38
rs771633923 Chr.18: 63655891-63655891 on Build GRCh38
rs577262694 Chr.18: 63655875-63655875 on Build GRCh38
rs770224229 Chr.18: 63655902-63655902 on Build GRCh38
rs755242648 Chr.18: 63656034-63656034 on Build GRCh38
rs146839299 Chr.18: 63655740-63655740 on Build GRCh38
rs757404837 Chr.18: 63655803-63655803 on Build GRCh38
rs751078614 Chr.18: 63656017-63656017 on Build GRCh38
rs780939570 Chr.18: 63656028-63656028 on Build GRCh38
rs748296283 Chr.18: 63655693-63655693 on Build GRCh38
rs768646518 Chr.18: 63655883-63655883 on Build GRCh38
rs577923279 Chr.18: 63655565-63655565 on Build GRCh38
rs563454281 Chr.18: 63655612-63655612 on Build GRCh38
rs201608695 Chr.18: 63655889-63655889 on Build GRCh38
rs1011826024 Chr.18: 63655873-63655873 on Build GRCh38
rs1047256 Chr.18: 63655660-63655660 on Build GRCh38
rs148202796 Chr.18: 63655830-63655830 on Build GRCh38
rs761199627 Chr.18: 63655708-63655708 on Build GRCh38
rs72132327 Chr.18: 63655778-63655779 on Build GRCh38
rs3180227 Chr.18: 63655778-63655778 on Build GRCh38
rs760049271 Chr.18: 63655640-63655640 on Build GRCh38
rs12953909 Chr.18: 63655746-63655746 on Build GRCh38
rs755428759 Chr.18: 63655755-63655755 on Build GRCh38
rs946134131 Chr.18: 63655623-63655623 on Build GRCh38
rs577782225 Chr.18: 63655812-63655812 on Build GRCh38
rs749066127 Chr.18: 63655829-63655829 on Build GRCh38
rs60533853 Chr.18: 63655777-63655778 on Build GRCh38
rs370963443 Chr.18: 63655948-63655948 on Build GRCh38
rs377088096 Chr.18: 63655937-63655937 on Build GRCh38
rs184759638 Chr.18: 63655792-63655792 on Build GRCh38
rs766672714 Chr.18: 63655611-63655612 on Build GRCh38
rs1004633937 Chr.18: 63655738-63655738 on Build GRCh38
rs565685500 Chr.18: 63655782-63655782 on Build GRCh38
rs375882446 Chr.18: 63655652-63655652 on Build GRCh38
rs143227753 Chr.18: 63655610-63655610 on Build GRCh38
rs149071083 Chr.18: 63655772-63655772 on Build GRCh38
rs752276065 Chr.18: 63655653-63655653 on Build GRCh38
rs374667283 Chr.18: 63655722-63655722 on Build GRCh38
rs570288715 Chr.18: 63655768-63655768 on Build GRCh38

View Full Product Details