Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002019.4 | NP_002010.2 | 17 - 18 | 2774 | 55 |
XM_017020485.1 | XP_016875974.1 | 17 - 18 | 2774 | 55 | |
GenBank mRNA | AB209050.1 | - | 1 - 2 | 130 | 55 |
AF063657.2 | - | 17 - 18 | 2489 | 55 | |
DQ836394.1 | - | 3 - 4 | 314 | 55 | |
DQ836395.1 | - | 4 - 5 | 376 | 55 | |
DQ836396.1 | - | 2 - 3 | 152 | 55 |