Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003384.2 | NP_003375.1 | 3 - 4 | 320 | 85 |
XM_006720247.3 | XP_006720310.1 | 3 - 4 | 358 | 85 | |
XM_011537132.1 | XP_011535434.1 | 2 - 3 | 219 | 85 | |
XM_017021622.1 | XP_016877111.1 | 2 - 3 | 219 | 85 | |
XM_017021624.1 | XP_016877113.1 | 3 - 4 | 358 | 85 | |
XM_017021625.1 | XP_016877114.1 | 3 - 4 | 399 | 85 | |
XM_017021626.1 | XP_016877115.1 | 3 - 4 | 358 | 85 | |
GenBank mRNA | AB000449.1 | - | 3 - 4 | 289 | 85 |
BC103761.1 | - | 3 - 4 | 298 | 85 | |
BC112075.1 | - | 3 - 4 | 288 | 85 | |
BC113510.1 | - | 3 - 4 | 288 | 85 | |
BU535261.1 | - | 3 - 4 | 340 | 85 |