Assay Design



Gene Transcripts

Gene Symbol
DGKD
Entrez Gene ID
8527
Gene Name
diacylglycerol kinase delta
Gene Aliases
DGKdelta, dgkd-2
Chromosome Location
Chr.2: 233354507 - 233472098 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003648.2 NP_003639.2 18 - 19 2322 77
NM_152879.2 NP_690618.2 19 - 20 2387 77
XM_005246107.3 XP_005246164.1 17 - 18 2054 77
XM_005246108.1 XP_005246165.1 17 - 18 2208 77
XM_005246110.3 XP_005246167.1 8 - 9 2059 77
XM_011512028.2 XP_011510330.1 19 - 20 3846 77
XM_011512029.2 XP_011510331.1 19 - 20 3847 77
XM_011512030.2 XP_011510332.1 19 - 20 2443 77
XM_011512031.2 XP_011510333.1 19 - 20 2317 77
XM_011512032.2 XP_011510334.1 19 - 20 2301 77
XM_011512033.2 XP_011510335.1 19 - 20 3848 77
XM_011512035.1 XP_011510337.1 17 - 18 2188 77
XM_011512037.2 XP_011510339.1 17 - 18 2292 77
XM_011512039.2 XP_011510341.1 19 - 20 3848 77
XM_011512040.2 XP_011510342.1 17 - 18 2224 77
XM_017005138.1 XP_016860627.1 19 - 20 2725 77
XM_017005139.1 XP_016860628.1 17 - 18 2217 77
XM_017005140.1 XP_016860629.1 19 - 20 2319 77
GenBank mRNA AB078966.1 - 19 - 20 2387 77
AK308846.1 - 17 - 18 2208 77
AK311044.1 - 17 - 18 2141 77
D63479.3 - 19 - 20 2328 77
D73409.1 - 18 - 19 2323 77

View Full Product Details