Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005108.3 | NP_005099.2 | 17 - 18 | 1551 | 64 |
XM_011534325.2 | XP_011532627.1 | 17 - 18 | 1532 | 64 | |
XM_011534327.1 | XP_011532629.1 | 17 - 18 | 1532 | 64 | |
XM_011534328.2 | XP_011532630.1 | 17 - 18 | 1532 | 64 | |
XM_011534329.1 | XP_011532631.1 | 17 - 18 | 1532 | 64 | |
XM_011534330.2 | XP_011532632.1 | 17 - 18 | 1532 | 64 | |
XM_017007595.1 | XP_016863084.1 | 15 - 16 | 1300 | 64 | |
XM_017007596.1 | XP_016863085.1 | 17 - 18 | 1577 | 64 | |
XM_017007597.1 | XP_016863086.1 | 9 - 10 | 866 | 64 | |
XM_017007598.1 | XP_016863087.1 | 17 - 18 | 1532 | 64 | |
XM_017007599.1 | XP_016863088.1 | 16 - 17 | 1435 | 64 | |
XM_017007600.1 | XP_016863089.1 | 14 - 15 | 1345 | 64 | |
GenBank mRNA | AB015046.1 | - | 17 - 18 | 1571 | 64 |
AK001205.1 | - | 10 - 11 | 822 | 64 | |
AK025728.1 | - | 17 - 18 | 1503 | 64 | |
AK293325.1 | - | 15 - 16 | 1305 | 64 | |
AK314386.1 | - | 17 - 18 | 1499 | 64 | |
BC137076.1 | - | 17 - 18 | 1483 | 64 | |
BC137080.1 | - | 17 - 18 | 1483 | 64 |