Assay Design



Gene Transcripts

Gene Symbol
SRC
Entrez Gene ID
6714
Gene Name
SRC proto-oncogene, non-receptor tyrosine kinase
Gene Aliases
ASV, SRC1, THC6, c-SRC, p60-Src
Chromosome Location
Chr.20: 37344685 - 37405432 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005417.4 NP_005408.1 7 - 8 1004 70
NM_198291.2 NP_938033.1 7 - 8 929 70
XM_011529013.2 XP_011527315.1 8 - 9 1273 70
XM_017028024.1 XP_016883513.1 9 - 10 1297 70
XM_017028025.1 XP_016883514.1 8 - 9 1178 70
XM_017028026.1 XP_016883515.1 8 - 9 1017 70
XM_017028027.1 XP_016883516.1 6 - 7 594 70
GenBank mRNA BC011566.1 - 5 - 6 594 70
BC051270.1 - 5 - 6 657 70

View Full Product Details