Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005813.4 | NP_005804.1 | 1 - 2 | 847 | 67 |
XM_005264237.3 | XP_005264294.1 | 2 - 3 | 1120 | 67 | |
XM_017003741.1 | XP_016859230.1 | 2 - 3 | 1070 | 67 | |
GenBank mRNA | AB015982.2 | - | 1 - 2 | 847 | 67 |
BC012820.2 | - | 1 - 2 | 806 | 67 | |
BC030706.2 | - | 1 - 2 | 792 | 67 |