Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000900.3 | NP_000891.2 | 1 - 2 | 186 | 108 |
GenBank mRNA | AK312029.1 | - | 1 - 2 | 121 | 108 |
AW997681.1 | - | 3 - 4 | 108 | ||
AY542304.1 | - | 1 - 2 | 69 | 108 | |
BC005272.1 | - | 1 - 2 | 121 | 108 | |
BC070314.1 | - | 1 - 2 | 94 | 108 | |
BT006733.1 | - | 1 - 2 | 58 | 108 | |
CB108937.1 | - | 1 - 2 | 153 | 108 | |
CR450358.1 | - | 1 - 2 | 58 | 108 | |
M58549.1 | - | 1 - 2 | 84 | 108 | |
X07362.1 | - | 1 - 2 | 64 | 108 | |
X53331.1 | - | 1 - 2 | 104 | 108 |