Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18785593-18786099 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COMP
Entrez Gene ID
1311
Gene Name
cartilage oligomeric matrix protein
Gene Aliases
EDM1, EPD1, MED, PSACH, THBS5, TSP5
UniGene
Hs.1584
Gene Chromosome Location
Chr.19: 18782773-18791305 on Build GRCh38
Transcripts
NM_000095.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5601300 c.1432C>T p.P478S Chr.19: 18786022-18786022 on Build GRCh38
310278 c.1424A>T p.D475V Chr.19: 18786030-18786030 on Build GRCh38
993159 c.1406A>G p.D469G Chr.19: 18786048-18786048 on Build GRCh38
3712861 c.1634C>T p.A545V Chr.19: 18785707-18785707 on Build GRCh38
417690 c.1475A>T p.Q492L Chr.19: 18785979-18785979 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs312262897 Chr.19: 18786035-18786037 on Build GRCh38
rs924124721 Chr.19: 18785936-18785936 on Build GRCh38
rs747845826 Chr.19: 18785806-18785806 on Build GRCh38
rs770721341 Chr.19: 18785973-18785975 on Build GRCh38
rs762683869 Chr.19: 18786013-18786013 on Build GRCh38
rs761831829 Chr.19: 18785940-18785940 on Build GRCh38
rs750363391 Chr.19: 18785773-18785773 on Build GRCh38
rs753646345 Chr.19: 18785863-18785863 on Build GRCh38
rs775902974 Chr.19: 18785915-18785915 on Build GRCh38
rs768778072 Chr.19: 18785920-18785920 on Build GRCh38
rs201977029 Chr.19: 18785895-18785895 on Build GRCh38
rs769866414 Chr.19: 18786008-18786008 on Build GRCh38
rs537092088 Chr.19: 18785880-18785880 on Build GRCh38
rs935596646 Chr.19: 18785741-18785741 on Build GRCh38
rs763122767 Chr.19: 18785757-18785757 on Build GRCh38
rs756726447 Chr.19: 18785968-18785968 on Build GRCh38
rs1056006024 Chr.19: 18785779-18785779 on Build GRCh38
rs554998204 Chr.19: 18785845-18785845 on Build GRCh38
rs1043943664 Chr.19: 18785696-18785696 on Build GRCh38
rs772268073 Chr.19: 18785987-18785987 on Build GRCh38
rs143714206 Chr.19: 18785966-18785966 on Build GRCh38
rs312262902 Chr.19: 18785762-18785762 on Build GRCh38
rs868493730 Chr.19: 18785997-18785997 on Build GRCh38
rs137852650 Chr.19: 18786040-18786040 on Build GRCh38
rs751036767 Chr.19: 18785967-18785967 on Build GRCh38
rs752438156 Chr.19: 18785959-18785959 on Build GRCh38
rs1056497456 Chr.19: 18785929-18785929 on Build GRCh38
rs369097923 Chr.19: 18785635-18785635 on Build GRCh38
rs747544637 Chr.19: 18785817-18785817 on Build GRCh38
rs780461514 Chr.19: 18786066-18786066 on Build GRCh38
rs145034923 Chr.19: 18785753-18785753 on Build GRCh38
rs373498615 Chr.19: 18785691-18785691 on Build GRCh38
rs137852651 Chr.19: 18786051-18786051 on Build GRCh38
rs757708852 Chr.19: 18785893-18785893 on Build GRCh38
rs758051214 Chr.19: 18785877-18785877 on Build GRCh38
rs773226781 Chr.19: 18785928-18785945 on Build GRCh38
rs772420147 Chr.19: 18785723-18785723 on Build GRCh38
rs760607070 Chr.19: 18785856-18785856 on Build GRCh38
rs762347570 Chr.19: 18785637-18785637 on Build GRCh38
rs776690209 Chr.19: 18785952-18785952 on Build GRCh38
rs778740025 Chr.19: 18785805-18785805 on Build GRCh38
rs312262903 Chr.19: 18785755-18785755 on Build GRCh38
rs1041411983 Chr.19: 18785662-18785662 on Build GRCh38
rs370767233 Chr.19: 18785914-18785914 on Build GRCh38
rs397515511 Chr.19: 18785676-18785676 on Build GRCh38
rs779933113 Chr.19: 18785786-18785786 on Build GRCh38
rs778270994 Chr.19: 18785866-18785867 on Build GRCh38
rs28933699 Chr.19: 18786043-18786043 on Build GRCh38
rs771590987 Chr.19: 18785807-18785807 on Build GRCh38
rs752778447 Chr.19: 18785672-18785672 on Build GRCh38
rs773490189 Chr.19: 18785991-18785991 on Build GRCh38
rs768883872 Chr.19: 18785984-18785984 on Build GRCh38
rs137852654 Chr.19: 18785772-18785772 on Build GRCh38
rs541996591 Chr.19: 18785983-18785983 on Build GRCh38
rs748950117 Chr.19: 18785648-18785648 on Build GRCh38
rs978668554 Chr.19: 18785615-18785615 on Build GRCh38
rs779812541 Chr.19: 18785637-18785637 on Build GRCh38
rs376180942 Chr.19: 18785909-18785909 on Build GRCh38
rs763986060 Chr.19: 18786021-18786021 on Build GRCh38
rs759687021 Chr.19: 18785751-18785751 on Build GRCh38
rs545882712 Chr.19: 18785901-18785901 on Build GRCh38
rs145283906 Chr.19: 18785737-18785737 on Build GRCh38
rs777269967 Chr.19: 18785878-18785878 on Build GRCh38
rs749463139 Chr.19: 18785919-18785919 on Build GRCh38
rs777205423 Chr.19: 18785814-18785814 on Build GRCh38
rs768270282 Chr.19: 18785630-18785630 on Build GRCh38
rs865949126 Chr.19: 18785998-18785998 on Build GRCh38
rs1043553569 Chr.19: 18785947-18785947 on Build GRCh38
rs758010826 Chr.19: 18785964-18785964 on Build GRCh38
rs761508334 Chr.19: 18786047-18786047 on Build GRCh38
rs749001252 Chr.19: 18785628-18785628 on Build GRCh38
rs750142844 Chr.19: 18785702-18785702 on Build GRCh38
rs200037349 Chr.19: 18785850-18785850 on Build GRCh38
rs746770208 Chr.19: 18785935-18785935 on Build GRCh38
rs34581246 Chr.19: 18785867-18785867 on Build GRCh38
rs377451339 Chr.19: 18785734-18785734 on Build GRCh38
rs999848388 Chr.19: 18785892-18785892 on Build GRCh38
rs373687591 Chr.19: 18785970-18785970 on Build GRCh38
rs916499370 Chr.19: 18785934-18785934 on Build GRCh38
rs747758496 Chr.19: 18785881-18785882 on Build GRCh38
rs200870935 Chr.19: 18785707-18785707 on Build GRCh38
rs746513458 Chr.19: 18785891-18785891 on Build GRCh38
rs759605845 Chr.19: 18785953-18785953 on Build GRCh38
rs769960335 Chr.19: 18785844-18785844 on Build GRCh38
rs751147399 Chr.19: 18786044-18786044 on Build GRCh38
rs773400537 Chr.19: 18785649-18785649 on Build GRCh38
rs577024872 Chr.19: 18785899-18785899 on Build GRCh38
rs763130585 Chr.19: 18785847-18785847 on Build GRCh38
rs868086369 Chr.19: 18785651-18785651 on Build GRCh38
rs765454754 Chr.19: 18785752-18785752 on Build GRCh38
rs777047473 Chr.19: 18785744-18785744 on Build GRCh38
rs756568697 Chr.19: 18786055-18786055 on Build GRCh38
rs765296978 Chr.19: 18785669-18785669 on Build GRCh38
rs375476152 Chr.19: 18786070-18786070 on Build GRCh38
rs867983497 Chr.19: 18785986-18785986 on Build GRCh38
rs918687726 Chr.19: 18785960-18785960 on Build GRCh38
rs951397973 Chr.19: 18785627-18785627 on Build GRCh38
rs768082784 Chr.19: 18785643-18785643 on Build GRCh38
rs761134568 Chr.19: 18785650-18785650 on Build GRCh38
rs1045068010 Chr.19: 18786009-18786009 on Build GRCh38
rs145290786 Chr.19: 18785653-18785653 on Build GRCh38
rs193922900 Chr.19: 18786035-18786037 on Build GRCh38
rs312262898 Chr.19: 18786034-18786035 on Build GRCh38
rs766442357 Chr.19: 18785859-18785859 on Build GRCh38
rs765174045 Chr.19: 18785869-18785869 on Build GRCh38
rs74432818 Chr.19: 18785660-18785660 on Build GRCh38
rs191645921 Chr.19: 18786001-18786001 on Build GRCh38
rs375299091 Chr.19: 18785624-18785624 on Build GRCh38
rs566112479 Chr.19: 18785709-18785709 on Build GRCh38
rs28936669 Chr.19: 18786036-18786036 on Build GRCh38
rs149083265 Chr.19: 18785823-18785823 on Build GRCh38
rs780411024 Chr.19: 18785971-18785971 on Build GRCh38
rs759913580 Chr.19: 18785835-18785835 on Build GRCh38
rs375031149 Chr.19: 18786050-18786050 on Build GRCh38
rs775306610 Chr.19: 18786012-18786012 on Build GRCh38
rs771885029 Chr.19: 18785919-18785919 on Build GRCh38
rs757080842 Chr.19: 18785685-18785685 on Build GRCh38
rs1001694143 Chr.19: 18785754-18785754 on Build GRCh38
rs761133261 Chr.19: 18785727-18785727 on Build GRCh38
rs766950514 Chr.19: 18786048-18786048 on Build GRCh38
rs780728105 Chr.19: 18785969-18785969 on Build GRCh38
rs951216686 Chr.19: 18785841-18785841 on Build GRCh38
rs535198633 Chr.19: 18785793-18785793 on Build GRCh38
rs765118743 Chr.19: 18785957-18785957 on Build GRCh38
rs779774621 Chr.19: 18785708-18785708 on Build GRCh38
rs774444113 Chr.19: 18785927-18785927 on Build GRCh38
rs756075225 Chr.19: 18785778-18785778 on Build GRCh38
rs747684271 Chr.19: 18785722-18785722 on Build GRCh38
rs754790637 Chr.19: 18785868-18785868 on Build GRCh38
rs754475799 Chr.19: 18785768-18785781 on Build GRCh38
rs1018789643 Chr.19: 18785942-18785942 on Build GRCh38
rs748431274 Chr.19: 18785985-18785985 on Build GRCh38
rs767746261 Chr.19: 18785941-18785941 on Build GRCh38
rs773740227 Chr.19: 18785724-18785724 on Build GRCh38
rs753703511 Chr.19: 18785790-18785790 on Build GRCh38

View Full Product Details