Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001139466.1 | NP_001132938.1 | 3 - 4 | 332 | 62 |
NM_005647.3 | NP_005638.1 | 3 - 4 | 451 | 62 | |
GenBank mRNA | AK290962.1 | - | 3 - 4 | 315 | 62 |
BC052304.1 | - | 3 - 4 | 451 | 62 | |
Y12781.1 | - | 3 - 4 | 610 | 62 |