Assay Design
Gene Transcripts
- Gene Symbol
- KCNK10
- Entrez Gene ID
-
54207
- Gene Name
- potassium two pore domain channel subfamily K member 10
- Gene Aliases
-
K2p10.1, PPP1R97, TREK-2, TREK2
- Chromosome Location
-
Chr.14: 88180108 - 88326912 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF279890.1 |
- |
1 - 2 |
511 |
146 |
AK315263.1 |
- |
1 - 2 |
164 |
146 |
EU978938.1 |
- |
1 - 2 |
325 |
146 |
View Full Product Details