Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000593.5 | NP_000584.2 | 10 - 11 | 2375 | 61 |
NM_001292022.1 | NP_001278951.1 | 10 - 11 | 1635 | 61 | |
GenBank mRNA | AB012644.1 | - | 10 - 11 | 2039 | 61 |
AK123494.1 | - | 4 - 5 | 1962 | 61 | |
AK130176.1 | - | 5 - 6 | 787 | 61 | |
AK302340.1 | - | 10 - 11 | 1606 | 61 | |
AK313762.1 | - | 10 - 11 | 2411 | 61 | |
AY523970.1 | - | 10 - 11 | 2246 | 61 | |
AY523971.2 | - | 10 - 11 | 2246 | 61 | |
BC014081.2 | - | 10 - 11 | 2375 | 61 | |
BX648013.1 | - | 10 - 11 | 2383 | 61 | |
KU178346.1 | - | 10 - 11 | 2220 | 61 | |
KU178347.1 | - | 4 - 5 | 765 | 61 | |
L21204.1 | - | 10 - 11 | 2040 | 61 | |
L21205.1 | - | 10 - 11 | 2040 | 61 | |
L21206.1 | - | 10 - 11 | 2040 | 61 | |
L21207.1 | - | 10 - 11 | 2040 | 61 | |
L21208.1 | - | 10 - 11 | 2040 | 61 | |
X57522.1 | - | 10 - 11 | 2250 | 61 |