Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015711.3 | NP_056526.3 | 11 - 12 | 3444 | 84 |
XM_005258833.4 | XP_005258890.1 | 11 - 12 | 3487 | 84 | |
XM_006723180.3 | XP_006723243.1 | 7 - 8 | 2407 | 84 | |
XM_011526882.2 | XP_011525184.1 | 7 - 8 | 4454 | 84 | |
XM_011526883.2 | XP_011525185.1 | 10 - 11 | 3310 | 84 | |
GenBank mRNA | AF182077.1 | - | 6 - 7 | 3460 | 84 |