Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006120.3 | NP_006111.2 | 1 - 2 | 175 | 100 |
GenBank mRNA | AK291858.1 | - | 1 - 2 | 111 | 100 |
BC011447.1 | - | 1 - 2 | 148 | 100 | |
BC026279.1 | - | 1 - 2 | 104 | 100 | |
CR450324.1 | - | 1 - 2 | 89 | 100 | |
X62744.1 | - | 1 - 2 | 134 | 100 |