Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 45549382-45549879 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OPA3
Entrez Gene ID
80207
Gene Name
optic atrophy 3 (autosomal recessive, with chorea and spastic paraplegia)
Gene Aliases
MGA3
UniGene
Hs.466945
Gene Chromosome Location
Chr.19: 45527767-45584864 on Build GRCh38
Transcripts
NM_001017989.2 NM_025136.3 XM_006723403.3

SNP Detail

SNP ID Chromosome Location
rs184285761 Chr.19: 45549449-45549449 on Build GRCh38
rs944182012 Chr.19: 45549625-45549625 on Build GRCh38
rs551539059 Chr.19: 45549655-45549655 on Build GRCh38
rs147972697 Chr.19: 45549542-45549542 on Build GRCh38
rs981647131 Chr.19: 45549760-45549760 on Build GRCh38
rs924482523 Chr.19: 45549543-45549543 on Build GRCh38
rs890552307 Chr.19: 45549710-45549710 on Build GRCh38
rs1051564521 Chr.19: 45549702-45549702 on Build GRCh38
rs959124917 Chr.19: 45549562-45549562 on Build GRCh38
rs780154471 Chr.19: 45549814-45549814 on Build GRCh38
rs993407622 Chr.19: 45549815-45549815 on Build GRCh38
rs7247946 Chr.19: 45549551-45549551 on Build GRCh38
rs868846381 Chr.19: 45549502-45549502 on Build GRCh38
rs534214560 Chr.19: 45549754-45549754 on Build GRCh38
rs1010295449 Chr.19: 45549727-45549727 on Build GRCh38
rs907521783 Chr.19: 45549637-45549637 on Build GRCh38
rs371821013 Chr.19: 45549438-45549438 on Build GRCh38
rs538573848 Chr.19: 45549430-45549430 on Build GRCh38
rs1020272145 Chr.19: 45549531-45549531 on Build GRCh38
rs546196266 Chr.19: 45549535-45549535 on Build GRCh38
rs971608620 Chr.19: 45549463-45549463 on Build GRCh38
rs62109650 Chr.19: 45549640-45549640 on Build GRCh38
rs191869562 Chr.19: 45549700-45549700 on Build GRCh38
rs914363561 Chr.19: 45549591-45549591 on Build GRCh38
rs555830952 Chr.19: 45549759-45549759 on Build GRCh38
rs74409501 Chr.19: 45549499-45549499 on Build GRCh38
rs1024454874 Chr.19: 45549845-45549845 on Build GRCh38
rs886054525 Chr.19: 45549669-45549669 on Build GRCh38
rs750504205 Chr.19: 45549779-45549779 on Build GRCh38
rs529245346 Chr.19: 45549544-45549544 on Build GRCh38
rs187028696 Chr.19: 45549630-45549630 on Build GRCh38
rs959439732 Chr.19: 45549433-45549433 on Build GRCh38
rs1010241697 Chr.19: 45549521-45549521 on Build GRCh38
rs990619244 Chr.19: 45549568-45549568 on Build GRCh38
rs932384787 Chr.19: 45549606-45549606 on Build GRCh38
rs71352273 Chr.19: 45549686-45549686 on Build GRCh38
rs1030822174 Chr.19: 45549519-45549519 on Build GRCh38
rs934350700 Chr.19: 45549563-45549563 on Build GRCh38
rs879119658 Chr.19: 45549487-45549488 on Build GRCh38
rs762519965 Chr.19: 45549440-45549440 on Build GRCh38
rs996301906 Chr.19: 45549460-45549460 on Build GRCh38
rs183951390 Chr.19: 45549804-45549804 on Build GRCh38
rs1049914310 Chr.19: 45549612-45549612 on Build GRCh38
rs541381652 Chr.19: 45549492-45549492 on Build GRCh38
rs968971001 Chr.19: 45549541-45549541 on Build GRCh38
rs989767343 Chr.19: 45549590-45549590 on Build GRCh38
rs886054524 Chr.19: 45549488-45549488 on Build GRCh38
rs1037512620 Chr.19: 45549418-45549418 on Build GRCh38
rs79660166 Chr.19: 45549636-45549636 on Build GRCh38
rs73568973 Chr.19: 45549792-45549792 on Build GRCh38
rs1026966922 Chr.19: 45549641-45549641 on Build GRCh38
rs900375467 Chr.19: 45549662-45549662 on Build GRCh38
rs1020324683 Chr.19: 45549728-45549728 on Build GRCh38
rs941929643 Chr.19: 45549412-45549412 on Build GRCh38
rs1022788544 Chr.19: 45549458-45549458 on Build GRCh38
rs757264789 Chr.19: 45549756-45549756 on Build GRCh38
rs895258547 Chr.19: 45549666-45549666 on Build GRCh38
rs752997579 Chr.19: 45549623-45549623 on Build GRCh38
rs200851409 Chr.19: 45549500-45549500 on Build GRCh38
rs890509236 Chr.19: 45549520-45549520 on Build GRCh38
rs369558782 Chr.19: 45549409-45549409 on Build GRCh38

View Full Product Details