Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271769.1 | NP_001258698.1 | 2 - 3 | 388 | 83 |
NM_003664.4 | NP_003655.3 | 2 - 3 | 376 | 83 | |
XM_005248618.3 | XP_005248675.1 | 2 - 3 | 348 | 83 | |
XM_005248619.4 | XP_005248676.1 | 2 - 3 | 348 | 83 | |
XM_017010001.1 | XP_016865490.1 | 2 - 3 | 128 | 83 | |
GenBank mRNA | AK225908.1 | - | 2 - 3 | 252 | 83 |
AK291201.1 | - | 2 - 3 | 317 | 83 | |
AL708147.1 | - | 2 - 3 | 338 | 83 | |
AY623424.1 | - | 2 - 3 | 278 | 83 | |
BC038444.1 | - | 2 - 3 | 337 | 83 | |
BX538041.1 | - | 2 - 3 | 326 | 83 | |
DA889915.1 | - | 2 - 3 | 376 | 83 | |
KU178426.1 | - | 2 - 3 | 200 | 83 | |
KU178427.1 | - | 2 - 3 | 200 | 83 | |
U81504.1 | - | 2 - 3 | 292 | 83 | |
U91931.1 | - | 2 - 3 | 253 | 83 |