Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003683.5 | NP_003674.1 | 5 - 6 | 519 | 75 |
XM_017028485.1 | XP_016883974.1 | 5 - 6 | 510 | 75 | |
GenBank mRNA | AK223185.1 | - | 5 - 6 | 480 | 75 |
AK223215.1 | - | 5 - 6 | 476 | 75 | |
AK303000.1 | - | 5 - 6 | 479 | 75 | |
AK313116.1 | - | 5 - 6 | 476 | 75 | |
AL137757.1 | - | 2 - 3 | 540 | 75 | |
BC000380.2 | - | 5 - 6 | 466 | 75 | |
BC014787.1 | - | 5 - 6 | 441 | 75 | |
BG829556.1 | - | - | 507 | 75 | |
U79775.1 | - | 5 - 6 | 464 | 75 |