Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 46747969-46748500 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FKRP
Entrez Gene ID
79147
Gene Name
fukutin related protein
Gene Aliases
LGMD2I, MDC1C, MDDGA5, MDDGB5, MDDGC5
UniGene
Hs.515493
Gene Chromosome Location
Chr.19: 46746015-46758575 on Build GRCh38
Transcripts
NM_001039885.2 NM_024301.4 XM_005259247.2 XM_005259248.2 XM_005259249.4 XM_011527306.1 XM_011527307.1 XM_017027297.1

SNP Detail

SNP ID Chromosome Location
rs941872245 Chr.19: 46748082-46748082 on Build GRCh38
rs927111508 Chr.19: 46748325-46748325 on Build GRCh38
rs974883370 Chr.19: 46748280-46748280 on Build GRCh38
rs541137919 Chr.19: 46748418-46748418 on Build GRCh38
rs896529678 Chr.19: 46748338-46748338 on Build GRCh38
rs368510249 Chr.19: 46748201-46748201 on Build GRCh38
rs980837948 Chr.19: 46748136-46748136 on Build GRCh38
rs182351921 Chr.19: 46748397-46748397 on Build GRCh38
rs960592657 Chr.19: 46748143-46748143 on Build GRCh38
rs919134301 Chr.19: 46748412-46748412 on Build GRCh38
rs529371486 Chr.19: 46748291-46748291 on Build GRCh38
rs998201287 Chr.19: 46748268-46748268 on Build GRCh38
rs111590642 Chr.19: 46748454-46748454 on Build GRCh38
rs866189992 Chr.19: 46748137-46748137 on Build GRCh38
rs367634015 Chr.19: 46748466-46748466 on Build GRCh38
rs780416928 Chr.19: 46748468-46748468 on Build GRCh38
rs181509391 Chr.19: 46748033-46748033 on Build GRCh38
rs1644332 Chr.19: 46748431-46748431 on Build GRCh38
rs1037464697 Chr.19: 46748115-46748115 on Build GRCh38
rs575662264 Chr.19: 46748435-46748435 on Build GRCh38
rs919186818 Chr.19: 46748208-46748208 on Build GRCh38
rs753849876 Chr.19: 46748219-46748219 on Build GRCh38
rs968188266 Chr.19: 46748332-46748332 on Build GRCh38
rs560682162 Chr.19: 46748281-46748281 on Build GRCh38
rs929105452 Chr.19: 46748218-46748218 on Build GRCh38
rs974452581 Chr.19: 46748016-46748016 on Build GRCh38
rs1037516168 Chr.19: 46748323-46748323 on Build GRCh38
rs907806227 Chr.19: 46748318-46748318 on Build GRCh38
rs1040093648 Chr.19: 46748241-46748241 on Build GRCh38
rs556023568 Chr.19: 46748074-46748074 on Build GRCh38
rs956832118 Chr.19: 46748071-46748071 on Build GRCh38
rs967863625 Chr.19: 46748121-46748121 on Build GRCh38
rs1022424508 Chr.19: 46748120-46748120 on Build GRCh38
rs571963158 Chr.19: 46748356-46748356 on Build GRCh38
rs1033864100 Chr.19: 46748376-46748376 on Build GRCh38
rs186004898 Chr.19: 46748151-46748151 on Build GRCh38
rs941924651 Chr.19: 46748319-46748319 on Build GRCh38
rs1014011628 Chr.19: 46748339-46748339 on Build GRCh38
rs28451715 Chr.19: 46748422-46748422 on Build GRCh38
rs189133927 Chr.19: 46748030-46748030 on Build GRCh38
rs1021626527 Chr.19: 46748220-46748220 on Build GRCh38
rs929042043 Chr.19: 46748407-46748407 on Build GRCh38
rs372268395 Chr.19: 46748334-46748334 on Build GRCh38
rs1002221682 Chr.19: 46748180-46748180 on Build GRCh38
rs1042362290 Chr.19: 46748371-46748371 on Build GRCh38
rs190514804 Chr.19: 46748313-46748313 on Build GRCh38
rs757040985 Chr.19: 46748240-46748240 on Build GRCh38
rs549415194 Chr.19: 46748292-46748292 on Build GRCh38
rs886315040 Chr.19: 46748058-46748058 on Build GRCh38
rs938138241 Chr.19: 46748328-46748328 on Build GRCh38
rs547680438 Chr.19: 46748393-46748393 on Build GRCh38
rs111868311 Chr.19: 46748025-46748025 on Build GRCh38
rs1032539223 Chr.19: 46748411-46748411 on Build GRCh38
rs573531526 Chr.19: 46748178-46748181 on Build GRCh38
rs750625805 Chr.19: 46748456-46748456 on Build GRCh38
rs562882514 Chr.19: 46748303-46748303 on Build GRCh38
rs1013877304 Chr.19: 46748209-46748209 on Build GRCh38
rs534454116 Chr.19: 46748385-46748385 on Build GRCh38
rs966916866 Chr.19: 46748221-46748221 on Build GRCh38
rs564816959 Chr.19: 46748140-46748140 on Build GRCh38
rs760557666 Chr.19: 46748001-46748001 on Build GRCh38
rs1032486927 Chr.19: 46748239-46748239 on Build GRCh38
rs1048100261 Chr.19: 46748056-46748056 on Build GRCh38
rs1009747399 Chr.19: 46748110-46748110 on Build GRCh38
rs1001045650 Chr.19: 46748238-46748238 on Build GRCh38
rs909073119 Chr.19: 46748423-46748423 on Build GRCh38
rs1055155213 Chr.19: 46748200-46748200 on Build GRCh38
rs964478192 Chr.19: 46748253-46748253 on Build GRCh38

View Full Product Details