Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004320.4 | NP_004311.1 | 1 - 2 | 301 | 71 |
NM_173201.3 | NP_775293.1 | 1 - 2 | 301 | 71 | |
GenBank mRNA | AK291314.1 | - | 1 - 2 | 230 | 71 |
BC037354.1 | - | 1 - 2 | 229 | 71 | |
BX537389.1 | - | 1 - 2 | 297 | 71 | |
BX537784.1 | - | 1 - 2 | 299 | 71 | |
BX647365.1 | - | 1 - 2 | 297 | 71 | |
BX647366.1 | - | 1 - 2 | 293 | 71 | |
BX647367.1 | - | 1 - 2 | 301 | 71 | |
BX648544.1 | - | 1 - 2 | 298 | 71 | |
BX649072.1 | - | 1 - 2 | 290 | 71 |