Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081563.2 | NP_001075032.1 | 7 - 8 | 1719 | 89 |
NM_001288764.1 | NP_001275693.1 | 9 - 10 | 1409 | 89 | |
NM_001288765.1 | NP_001275694.1 | 6 - 7 | 990 | 89 | |
NM_004409.4 | NP_004400.4 | 8 - 9 | 1350 | 89 | |
GenBank mRNA | AK297299.1 | - | 6 - 7 | 941 | 89 |
BC026328.1 | - | 9 - 10 | 1404 | 89 | |
BC062553.1 | - | 8 - 9 | 1271 | 89 | |
KJ534826.1 | - | 7 - 8 | 1175 | 89 | |
KJ534827.1 | - | 7 - 8 | 1175 | 89 | |
M87312.1 | - | 7 - 8 | 1720 | 89 |