Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008844.2 | NP_001008844.1 | 4 - 5 | 941 | 74 |
NM_001319034.1 | NP_001305963.1 | 4 - 5 | 941 | 74 | |
NM_004415.3 | NP_004406.2 | 4 - 5 | 941 | 74 | |
GenBank mRNA | AB209992.1 | - | 4 - 5 | 899 | 74 |
AK296756.1 | - | 2 - 3 | 143 | 74 | |
AK301676.1 | - | 2 - 3 | 124 | 74 | |
AK304182.1 | - | 4 - 5 | 844 | 74 | |
BC140802.1 | - | 4 - 5 | 874 | 74 | |
HM151899.1 | - | 4 - 5 | 879 | 74 | |
M77830.3 | - | 4 - 5 | 879 | 74 |