Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166159.1 | NP_001159631.1 | 3 - 4 | 632 | 80 |
NM_004550.4 | NP_004541.1 | 4 - 5 | 841 | 80 | |
XM_005245208.2 | XP_005245265.1 | 4 - 5 | 743 | 80 | |
XM_005245209.1 | XP_005245266.1 | 2 - 3 | 312 | 80 | |
XM_017001357.1 | XP_016856846.1 | 3 - 4 | 1107 | 80 | |
GenBank mRNA | AF013160.1 | - | 3 - 4 | 399 | 80 |
AF050640.1 | - | 3 - 4 | 399 | 80 | |
AK222618.1 | - | 3 - 4 | 398 | 80 | |
AK301641.1 | - | 3 - 4 | 446 | 80 | |
AK314807.1 | - | 3 - 4 | 405 | 80 | |
AK315977.1 | - | 2 - 3 | 300 | 80 | |
BC000170.2 | - | 3 - 4 | 400 | 80 | |
BC001456.1 | - | 3 - 4 | 394 | 80 | |
BC008868.2 | - | 4 - 5 | 841 | 80 |