Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004654.3 | NP_004645.2 | 15 - 16 | 2843 | 71 |
XM_017030078.1 | XP_016885567.1 | 14 - 15 | 1998 | 71 | |
GenBank mRNA | AF000986.2 | - | 15 - 16 | 2843 | 71 |
Y13618.1 | - | 13 - 14 | 1955 | 71 | |
Y13619.1 | - | 13 - 14 | 1955 | 71 |