Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004724.3 | NP_004715.1 | 15 - 16 | 2317 | 101 |
XM_017018558.1 | XP_016874047.1 | 14 - 15 | 2065 | 101 | |
XM_017018559.1 | XP_016874048.1 | 15 - 16 | 2273 | 101 | |
GenBank mRNA | AK303871.1 | - | 14 - 15 | 1996 | 101 |
AK312925.1 | - | 15 - 16 | 2295 | 101 | |
BC128264.1 | - | 15 - 16 | 2196 | 101 | |
BQ773926.1 | - | 1 - 2 | 101 | ||
U54996.1 | - | 15 - 16 | 2243 | 101 |