Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256672.1 | NP_001243601.1 | 4 - 5 | 721 | 63 |
NM_001256673.1 | NP_001243602.1 | 3 - 4 | 571 | 63 | |
NM_001256674.1 | NP_001243603.1 | 3 - 4 | 571 | 63 | |
NM_001256675.1 | NP_001243604.1 | 4 - 5 | 721 | 63 | |
NM_001256676.1 | NP_001243605.1 | 4 - 5 | 535 | 63 | |
NM_001256677.1 | NP_001243606.1 | 5 - 6 | 852 | 63 | |
NM_001324226.1 | NP_001311155.1 | 3 - 4 | 840 | 63 | |
NM_001324227.1 | NP_001311156.1 | 4 - 5 | 535 | 63 | |
NM_001324228.1 | NP_001311157.1 | 4 - 5 | 599 | 63 | |
NM_001324229.1 | NP_001311158.1 | 3 - 4 | 1026 | 63 | |
NM_001324230.1 | NP_001311159.1 | 5 - 6 | 852 | 63 | |
NM_004809.4 | NP_004800.2 | 4 - 5 | 721 | 63 | |
XM_005254792.4 | XP_005254849.1 | 4 - 5 | 1625 | 63 | |
XM_006720773.3 | XP_006720836.1 | 4 - 5 | 1626 | 63 | |
GenBank mRNA | AF074953.1 | - | 4 - 5 | 64 | |
AF111800.1 | - | 3 - 4 | 520 | 63 | |
AF156557.1 | - | 4 - 5 | 729 | 63 | |
AK222665.1 | - | 4 - 5 | 604 | 63 | |
AK300799.1 | - | 5 - 6 | 736 | 63 | |
AK302051.1 | - | 3 - 4 | 526 | 63 | |
AL109664.1 | - | 3 - 4 | 401 | 63 | |
AL109665.1 | - | 4 - 5 | 575 | 63 | |
AL533460.3 | - | 4 - 5 | 512 | 63 | |
BC034379.1 | - | 4 - 5 | 667 | 63 | |
BC065249.1 | - | 4 - 5 | 667 | 63 | |
BX464941.2 | - | 4 - 5 | 535 | 63 | |
CR542089.1 | - | 4 - 5 | 596 | 63 | |
DQ064605.1 | - | 4 - 5 | 596 | 63 | |
Y16522.1 | - | 4 - 5 | 640 | 63 |