Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004911.4 | NP_004902.1 | 4 - 5 | 899 | 102 |
XM_006716185.2 | XP_006716248.1 | 4 - 5 | 899 | 102 | |
GenBank mRNA | AK290971.1 | - | 4 - 5 | 684 | 102 |
AK313299.1 | - | 4 - 5 | 102 | ||
BC000425.2 | - | 4 - 5 | 661 | 102 | |
BC001928.1 | - | 4 - 5 | 661 | 102 | |
BC006344.2 | - | 4 - 5 | 849 | 102 | |
BC011754.2 | - | 4 - 5 | 850 | 102 |