Assay Design
Gene Transcripts
- Gene Symbol
- KCNJ9
- Entrez Gene ID
-
3765
- Gene Name
- potassium voltage-gated channel subfamily J member 9
- Gene Aliases
-
GIRK3, KIR3.3
- Chromosome Location
-
Chr.1: 160055001 - 160090563 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_004983.2 |
NP_004974.2 |
1 - 2 |
129 |
72 |
GenBank mRNA |
U52152.1 |
- |
1 - 2 |
145 |
72 |
View Full Product Details