Assay Design



Gene Transcripts

Gene Symbol
MLF2
Entrez Gene ID
8079
Gene Name
myeloid leukemia factor 2
Gene Aliases
NTN4
Chromosome Location
Chr.12: 6747992 - 6753470 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_026581.1 1 - 2 530 64
GenBank mRNA AF070539.1 - 1 - 2 163 64
BC000898.2 - 1 - 2 103 64
BC002340.2 - 1 - 2 143 64
BI544309.1 - 1 - 2 536 64
U57342.1 - 1 - 2 149 64

Gene Symbol
TRIM54
Entrez Gene ID
57159
Gene Name
tripartite motif containing 54
Gene Aliases
MURF, MURF-3, RNF30, muRF3
Chromosome Location
Chr.2: 27282429 - 27307439 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK300699.1 - - 1368 64

View Full Product Details