Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005534.3 | NP_005525.2 | 3 - 4 | 1061 | 92 |
XM_005260969.2 | XP_005261026.1 | 4 - 5 | 577 | 92 | |
XM_011529553.1 | XP_011527855.1 | 4 - 5 | 503 | 92 | |
XM_011529554.2 | XP_011527856.1 | 3 - 4 | 937 | 92 | |
GenBank mRNA | AK292246.1 | - | 3 - 4 | 509 | 92 |
AK314005.1 | - | 3 - 4 | 522 | 92 | |
BC003624.2 | - | 3 - 4 | 529 | 92 | |
CB992729.1 | - | 3 - 4 | 571 | 92 | |
U05875.1 | - | 3 - 4 | 1061 | 92 | |
U05877.1 | - | 3 - 4 | 526 | 92 |