Assay Design



Gene Transcripts

Gene Symbol
USPL1
Entrez Gene ID
10208
Gene Name
ubiquitin specific peptidase like 1
Gene Aliases
C13orf22, D13S106E, bA121O19.1
Chromosome Location
Chr.13: 30617397 - 30660770 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005800.4 NP_005791.3 1 - 2 363 106
XM_006719751.3 XP_006719814.1 1 - 2 659 106
XM_017020314.1 XP_016875803.1 1 - 2 659 106
GenBank mRNA AK289826.1 - 1 - 2 119 106
BC036027.1 - 1 - 2 135 106
BC038103.2 - 1 - 2 273 106
X59131.1 - 1 - 2 108 106

View Full Product Details