Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037537.1 | NP_001032626.1 | 3 - 4 | 610 | 97 |
NM_001323080.1 | NP_001310009.1 | 4 - 5 | 669 | 97 | |
NM_001323082.1 | NP_001310011.1 | 4 - 5 | 505 | 97 | |
NM_001323084.1 | NP_001310013.1 | 3 - 4 | 610 | 97 | |
NM_006214.3 | NP_006205.1 | 4 - 5 | 505 | 97 | |
GenBank mRNA | AF023462.1 | - | 4 - 5 | 444 | 97 |
AF112977.1 | - | 4 - 5 | 476 | 97 | |
BC029512.1 | - | 4 - 5 | 465 | 97 | |
BF509272.1 | - | 3 - 4 | 627 | 97 | |
CR542055.1 | - | 4 - 5 | 417 | 97 |