Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170750.1 | NP_001164221.1 | 2 - 3 | 424 | 61 |
NM_001170752.1 | NP_001164223.1 | 3 - 4 | 484 | 61 | |
NM_006307.4 | NP_006298.1 | 3 - 4 | 484 | 61 | |
XM_017029893.1 | XP_016885382.1 | 3 - 4 | 423 | 61 | |
GenBank mRNA | AK125542.1 | - | 2 - 3 | 405 | 61 |
AK289430.1 | - | 3 - 4 | 431 | 61 | |
AK293127.1 | - | 3 - 4 | 461 | 61 | |
BC020684.1 | - | 3 - 4 | 439 | 61 | |
DA422854.1 | - | 2 - 3 | 397 | 61 | |
DC388035.1 | - | 3 - 4 | 484 | 61 | |
EU668359.1 | - | 3 - 4 | 390 | 61 | |
U61374.1 | - | 3 - 4 | 390 | 61 | |
U78093.1 | - | 3 - 4 | 391 | 61 |