Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286277.1 | NP_001273206.1 | 13 - 14 | 1729 | 70 |
NM_001286278.1 | NP_001273207.1 | 13 - 14 | 1759 | 70 | |
NM_006322.5 | NP_006313.1 | 13 - 14 | 1759 | 70 | |
XM_005268293.1 | XP_005268350.1 | 13 - 14 | 1754 | 70 | |
XM_011537462.2 | XP_011535764.1 | 11 - 12 | 1412 | 70 | |
XM_011537463.2 | XP_011535765.1 | 14 - 15 | 1688 | 70 | |
XM_011537464.2 | XP_011535766.1 | 13 - 14 | 2492 | 70 | |
XM_017020323.1 | XP_016875812.1 | 14 - 15 | 2106 | 70 | |
GenBank mRNA | AF042378.1 | - | 13 - 14 | 1638 | 70 |
AJ003061.1 | - | 13 - 14 | 1554 | 70 | |
AJ003062.1 | - | 13 - 14 | 1554 | 70 | |
AK302537.1 | - | 13 - 14 | 1676 | 70 | |
AK314785.1 | - | 13 - 14 | 1759 | 70 | |
BC013781.1 | - | 13 - 14 | 1699 | 70 | |
BC046634.2 | - | 13 - 14 | 1742 | 70 | |
BT009772.1 | - | 13 - 14 | 1554 | 70 |