Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001248006.1 | NP_001234935.1 | 2 - 3 | 522 | 71 |
NM_006458.3 | NP_006449.2 | 3 - 4 | 523 | 71 | |
NM_033278.3 | NP_150594.2 | 2 - 3 | 358 | 71 | |
XM_011519844.1 | XP_011518146.1 | 3 - 4 | 622 | 71 | |
XM_011519845.2 | XP_011518147.1 | 2 - 3 | 308 | 71 | |
XM_017017098.1 | XP_016872587.1 | 2 - 3 | 334 | 71 | |
XM_017017099.1 | XP_016872588.1 | 2 - 3 | 231 | 71 | |
GenBank mRNA | AF045239.1 | - | 1 - 2 | 127 | 71 |
AF220020.1 | - | 1 - 2 | 127 | 71 | |
AF220021.1 | - | 1 - 2 | 127 | 71 | |
AF220022.1 | - | 1 - 2 | 127 | 71 | |
AK126685.1 | - | 2 - 3 | 237 | 71 | |
AK291396.1 | - | 2 - 3 | 522 | 71 | |
AK299586.1 | - | 2 - 3 | 315 | 71 | |
AK311127.1 | - | 2 - 3 | 318 | 71 | |
AL360192.1 | - | 1 - 2 | 102 | 71 | |
BC096827.1 | - | 2 - 3 | 358 | 71 | |
BI916995.1 | - | 3 - 4 | 462 | 71 | |
BU500178.1 | - | - | 368 | 71 | |
DA089323.1 | - | 2 - 3 | 233 | 71 |