Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006998.3 | NP_008929.2 | 3 - 4 | 447 | 84 |
XM_017010181.1 | XP_016865670.1 | 2 - 3 | 159 | 84 | |
GenBank mRNA | AF420280.1 | - | 2 - 3 | 173 | 84 |
AK289477.1 | - | 3 - 4 | 502 | 84 | |
BC000336.2 | - | 3 - 4 | 414 | 84 | |
BC003036.1 | - | 3 - 4 | 414 | 84 | |
BM511122.1 | - | 3 - 4 | 453 | 84 | |
Y16752.1 | - | 3 - 4 | 400 | 84 |