Assay Design



Gene Transcripts

Gene Symbol
ZPBP
Entrez Gene ID
11055
Gene Name
zona pellucida binding protein
Gene Aliases
ZPBP1
Chromosome Location
Chr.7: 49846355 - 50093264 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159878.1 NP_001153350.1 2 - 3 279 67
NM_007009.2 NP_008940.2 2 - 3 279 67
XM_011515095.2 XP_011513397.1 2 - 3 273 67
XM_011515096.2 XP_011513398.1 2 - 3 251 67
XM_011515097.1 XP_011513399.1 2 - 3 273 67
XM_011515101.2 XP_011513403.1 2 - 3 227 67
XM_011515102.2 XP_011513404.1 2 - 3 452 67
XM_011515103.1 XP_011513405.1 2 - 3 273 67
XM_017011707.1 XP_016867196.1 2 - 3 273 67
GenBank mRNA AK312799.1 - 2 - 3 251 67
BC005223.1 - 2 - 3 238 67
BT006822.1 - 2 - 3 209 67
D17570.1 - 2 - 3 245 67

View Full Product Details