Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 166198772-166199271 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC101929680
Entrez Gene ID
101929680
Gene Name
uncharacterized LOC101929680
Gene Aliases
-
UniGene
Hs.662210
Gene Chromosome Location
Chr.2: 166081531-166301784 on Build GRCh38
Transcripts
NR_110260.1 NM_002977.3 XM_005246757.2 XM_011511616.2 XM_011511617.2 XM_011511618.2 XM_017004668.1 XM_017004669.1

Gene Symbol
SCN9A
Entrez Gene ID
6335
Gene Name
sodium voltage-gated channel alpha subunit 9
Gene Aliases
ETHA, FEB3B, GEFSP7, HSAN2D, NE-NA, NENA, Nav1.7, PN1, SFNP
UniGene
Hs.439145
Gene Chromosome Location
Chr.2: 166195185-166375987 on Build GRCh38
Transcripts
NR_110260.1 NM_002977.3 XM_005246757.2 XM_011511616.2 XM_011511617.2 XM_011511618.2 XM_017004668.1 XM_017004669.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5961959 c.5678G>A p.R1893H Chr.2: 166198928-166198928 on Build GRCh38
5394777 c.5467C>T p.Q1823* Chr.2: 166199175-166199175 on Build GRCh38
3406998 c.5482G>T p.D1828Y Chr.2: 166199160-166199160 on Build GRCh38
3569139 c.5625G>A p.R1875R Chr.2: 166198981-166198981 on Build GRCh38
4819867 c.5773G>A p.D1925N Chr.2: 166198869-166198869 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759757550 Chr.2: 166198919-166198919 on Build GRCh38
rs751325279 Chr.2: 166199152-166199152 on Build GRCh38
rs180949263 Chr.2: 166198934-166198934 on Build GRCh38
rs879254298 Chr.2: 166199024-166199024 on Build GRCh38
rs761344025 Chr.2: 166199049-166199049 on Build GRCh38
rs200876442 Chr.2: 166198889-166198889 on Build GRCh38
rs750503411 Chr.2: 166199196-166199196 on Build GRCh38
rs540262433 Chr.2: 166198955-166198955 on Build GRCh38
rs1021905006 Chr.2: 166198844-166198844 on Build GRCh38
rs200206979 Chr.2: 166198933-166198933 on Build GRCh38
rs202048301 Chr.2: 166198864-166198864 on Build GRCh38
rs560168893 Chr.2: 166198983-166198983 on Build GRCh38
rs372083483 Chr.2: 166198929-166198929 on Build GRCh38
rs779947651 Chr.2: 166198831-166198833 on Build GRCh38
rs781049315 Chr.2: 166199073-166199073 on Build GRCh38
rs759259640 Chr.2: 166199165-166199165 on Build GRCh38
rs770416478 Chr.2: 166199247-166199247 on Build GRCh38
rs763201215 Chr.2: 166198901-166198901 on Build GRCh38
rs752262872 Chr.2: 166198824-166198824 on Build GRCh38
rs201561928 Chr.2: 166199051-166199051 on Build GRCh38
rs748702835 Chr.2: 166199236-166199236 on Build GRCh38
rs772025034 Chr.2: 166198824-166198829 on Build GRCh38
rs201972510 Chr.2: 166199166-166199166 on Build GRCh38
rs377022631 Chr.2: 166198810-166198810 on Build GRCh38
rs201638678 Chr.2: 166199142-166199142 on Build GRCh38
rs776271263 Chr.2: 166199024-166199024 on Build GRCh38
rs746119070 Chr.2: 166199116-166199116 on Build GRCh38
rs775777109 Chr.2: 166199127-166199127 on Build GRCh38
rs558053149 Chr.2: 166198946-166198946 on Build GRCh38
rs757285775 Chr.2: 166199087-166199087 on Build GRCh38
rs772061182 Chr.2: 166198888-166198888 on Build GRCh38
rs200745478 Chr.2: 166199246-166199246 on Build GRCh38
rs367719041 Chr.2: 166198975-166198975 on Build GRCh38
rs201796657 Chr.2: 166198940-166198940 on Build GRCh38
rs200478305 Chr.2: 166199008-166199008 on Build GRCh38
rs747340707 Chr.2: 166199131-166199131 on Build GRCh38
rs778928947 Chr.2: 166199183-166199183 on Build GRCh38
rs746965222 Chr.2: 166198964-166198964 on Build GRCh38
rs895014544 Chr.2: 166198989-166198989 on Build GRCh38
rs201035087 Chr.2: 166198972-166198972 on Build GRCh38
rs754924014 Chr.2: 166199072-166199072 on Build GRCh38
rs764685430 Chr.2: 166199025-166199025 on Build GRCh38
rs201970521 Chr.2: 166198891-166198891 on Build GRCh38
rs776299330 Chr.2: 166199037-166199037 on Build GRCh38
rs200252925 Chr.2: 166198967-166198967 on Build GRCh38
rs945489250 Chr.2: 166199230-166199231 on Build GRCh38
rs768125863 Chr.2: 166198900-166198904 on Build GRCh38
rs371379751 Chr.2: 166199112-166199112 on Build GRCh38
rs577823520 Chr.2: 166198947-166198947 on Build GRCh38
rs984887572 Chr.2: 166198875-166198875 on Build GRCh38
rs371656473 Chr.2: 166198873-166198873 on Build GRCh38
rs756943130 Chr.2: 166198866-166198866 on Build GRCh38
rs200196731 Chr.2: 166199148-166199148 on Build GRCh38
rs774488202 Chr.2: 166198982-166198982 on Build GRCh38
rs779790941 Chr.2: 166198890-166198890 on Build GRCh38
rs3750904 Chr.2: 166198883-166198883 on Build GRCh38
rs876661248 Chr.2: 166199076-166199076 on Build GRCh38
rs762575742 Chr.2: 166199151-166199151 on Build GRCh38
rs1021711301 Chr.2: 166199181-166199181 on Build GRCh38
rs769295581 Chr.2: 166199135-166199135 on Build GRCh38
rs749373098 Chr.2: 166199077-166199077 on Build GRCh38
rs199954555 Chr.2: 166199214-166199214 on Build GRCh38
rs755510753 Chr.2: 166198833-166198833 on Build GRCh38
rs1012108087 Chr.2: 166199179-166199179 on Build GRCh38
rs756016813 Chr.2: 166199180-166199180 on Build GRCh38
rs773252059 Chr.2: 166198894-166198894 on Build GRCh38
rs200024315 Chr.2: 166198870-166198870 on Build GRCh38
rs774751626 Chr.2: 166198918-166198918 on Build GRCh38
rs371454107 Chr.2: 166198819-166198819 on Build GRCh38
rs760097843 Chr.2: 166198802-166198802 on Build GRCh38
rs1026656279 Chr.2: 166199098-166199098 on Build GRCh38
rs910321137 Chr.2: 166198898-166198898 on Build GRCh38
rs752580340 Chr.2: 166199172-166199172 on Build GRCh38
rs867949537 Chr.2: 166199038-166199038 on Build GRCh38
rs761153502 Chr.2: 166199016-166199016 on Build GRCh38
rs910761183 Chr.2: 166198877-166198877 on Build GRCh38
rs772335195 Chr.2: 166199118-166199118 on Build GRCh38
rs199572382 Chr.2: 166199053-166199053 on Build GRCh38
rs746685972 Chr.2: 166198841-166198842 on Build GRCh38
rs201483184 Chr.2: 166199084-166199084 on Build GRCh38
rs79805025 Chr.2: 166198928-166198928 on Build GRCh38
rs373922665 Chr.2: 166198881-166198881 on Build GRCh38
rs768070346 Chr.2: 166198926-166198926 on Build GRCh38
rs754909677 Chr.2: 166198966-166198966 on Build GRCh38
rs372311962 Chr.2: 166199231-166199231 on Build GRCh38
rs772373532 Chr.2: 166198986-166198986 on Build GRCh38
rs529045323 Chr.2: 166199009-166199009 on Build GRCh38
rs199705100 Chr.2: 166199096-166199096 on Build GRCh38
rs200803311 Chr.2: 166199149-166199149 on Build GRCh38
rs879254324 Chr.2: 166199219-166199219 on Build GRCh38
rs751361718 Chr.2: 166199069-166199069 on Build GRCh38
rs761434362 Chr.2: 166199141-166199141 on Build GRCh38
rs747367937 Chr.2: 166199227-166199227 on Build GRCh38
rs876661011 Chr.2: 166199078-166199078 on Build GRCh38
rs771138215 Chr.2: 166198910-166198910 on Build GRCh38
rs769712540 Chr.2: 166198893-166198893 on Build GRCh38
rs111558968 Chr.2: 166198860-166198860 on Build GRCh38
rs767893173 Chr.2: 166198804-166198804 on Build GRCh38
rs766124857 Chr.2: 166199060-166199060 on Build GRCh38
rs986172214 Chr.2: 166198892-166198892 on Build GRCh38
rs200608140 Chr.2: 166199136-166199136 on Build GRCh38
rs769097206 Chr.2: 166199229-166199229 on Build GRCh38
rs866918452 Chr.2: 166198874-166198874 on Build GRCh38
rs200956485 Chr.2: 166198935-166198935 on Build GRCh38
rs200855931 Chr.2: 166198863-166198863 on Build GRCh38
rs745699120 Chr.2: 166198809-166198809 on Build GRCh38
rs202239457 Chr.2: 166198973-166198973 on Build GRCh38
rs779128646 Chr.2: 166199114-166199114 on Build GRCh38
rs200410805 Chr.2: 166198848-166198848 on Build GRCh38
rs200494490 Chr.2: 166199061-166199061 on Build GRCh38

View Full Product Details