Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164392.1 | NP_001157864.1 | 1 - 2 | 296 | 67 |
NM_007358.3 | NP_031384.1 | 1 - 2 | 296 | 67 | |
XM_011541016.1 | XP_011539318.1 | 1 - 2 | 248 | 67 | |
XM_011541017.2 | XP_011539319.1 | 1 - 2 | 312 | 67 | |
XM_017000677.1 | XP_016856166.1 | 1 - 2 | 248 | 67 | |
XM_017000679.1 | XP_016856168.1 | 1 - 2 | 248 | 67 | |
GenBank mRNA | AF086908.1 | - | 1 - 2 | 221 | 67 |
AJ010014.1 | - | 1 - 2 | 247 | 67 | |
AK290318.1 | - | 1 - 2 | 248 | 67 | |
BC001897.1 | - | 1 - 2 | 215 | 67 | |
BC010013.2 | - | 1 - 2 | 220 | 67 | |
DB173315.1 | - | 1 - 2 | 296 | 67 |