Assay Design



Gene Transcripts

Gene Symbol
IL36RN
Entrez Gene ID
26525
Gene Name
interleukin 36 receptor antagonist
Gene Aliases
FIL1, FIL1(DELTA), FIL1D, IL-36Ra, IL1F5, IL1HY1, IL1L1, IL1RP3, IL36RA, PSORP, PSORS14
Chromosome Location
Chr.2: 113058638 - 113064744 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012275.2 NP_036407.1 2 - 3 185 116
NM_173170.1 NP_775262.1 2 - 3 157 116
GenBank mRNA AF186094.1 - 2 - 3 186 116
AF201830.1 - 1 - 2 51 116
AK290249.1 - 2 - 3 95 116
AY359117.1 - 2 - 3 90 116
BC024747.1 - 2 - 3 157 116

View Full Product Details