Assay Design



Gene Transcripts

Gene Symbol
MYLIP
Entrez Gene ID
29116
Gene Name
myosin regulatory light chain interacting protein
Gene Aliases
IDOL, MIR
Chromosome Location
Chr.6: 16129086 - 16151015 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013262.3 NP_037394.2 2 - 3 514 75
XM_005249032.2 XP_005249089.1 2 - 3 514 75
XM_017010789.1 XP_016866278.1 2 - 3 514 75
GenBank mRNA AF006003.1 - 2 - 3 427 75
AF187016.2 - 2 - 3 451 75
AF258586.1 - 2 - 3 491 75
AK074391.1 - 2 - 3 514 75
AK314648.1 - 2 - 3 514 75
BC002860.2 - 2 - 3 495 75
BT007055.1 - 2 - 3 277 75

View Full Product Details