Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013293.4 | NP_037425.1 | 1 - 2 | 255 | 84 |
XM_011515331.1 | XP_011513633.1 | 1 - 2 | 246 | 84 | |
GenBank mRNA | AB097048.1 | - | 1 - 2 | 196 | 84 |
AK314801.1 | - | 1 - 2 | 196 | 84 | |
BC017094.2 | - | 1 - 2 | 166 | 84 | |
BG722500.1 | - | 1 - 2 | 259 | 84 | |
BP376263.1 | - | 1 - 2 | 246 | 84 | |
U53209.1 | - | 1 - 2 | 195 | 84 |