Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013351.1 | NP_037483.1 | 1 - 2 | 707 | 62 |
GenBank mRNA | AF093098.1 | - | 1 - 2 | 707 | 62 |
AF241243.2 | - | 1 - 2 | 542 | 62 | |
AK223634.1 | - | 1 - 2 | 700 | 62 | |
BC039739.2 | - | 1 - 2 | 672 | 62 |