Assay Design



Gene Transcripts

Gene Symbol
FOXP3
Entrez Gene ID
50943
Gene Name
forkhead box P3
Gene Aliases
AIID, DIETER, IPEX, JM2, PIDX, XPID
Chromosome Location
Chr.X: 49250436 - 49266505 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114377.1 NP_001107849.1 9 - 10 1132 64
NM_014009.3 NP_054728.2 10 - 11 1237 64
XM_006724533.2 XP_006724596.2 9 - 10 1412 64
XM_011543916.2 XP_011542218.1 10 - 11 2662 64
XM_017029565.1 XP_016885054.1 11 - 12 2698 64
XM_017029566.1 XP_016885055.1 10 - 11 2662 64
XM_017029567.1 XP_016885056.1 9 - 10 1057 64
GenBank mRNA AF277993.1 - 10 - 11 1237 64
AJ005891.1 - 9 - 10 899 64
AK299988.1 - 9 - 10 1016 64
BC113401.1 - 9 - 10 1079 64
BC113403.1 - 9 - 10 1079 64
BC143785.1 - 8 - 9 974 64
BC143786.1 - 8 - 9 1148 64
DQ010327.1 - 8 - 9 944 64
EF534714.1 - 9 - 10 1049 64
EU855812.1 - 8 - 9 968 64

View Full Product Details