Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001287223.1 | NP_001274152.1 | 13 - 14 | 1872 | 93 |
NM_014139.2 | NP_054858.2 | 12 - 13 | 2041 | 93 | |
XM_011533321.2 | XP_011531623.1 | 9 - 10 | 1266 | 93 | |
XM_017005647.1 | XP_016861136.1 | 17 - 18 | 2493 | 93 | |
XM_017005648.1 | XP_016861137.1 | 14 - 15 | 1974 | 93 | |
XM_017005649.1 | XP_016861138.1 | 16 - 17 | 2371 | 93 | |
XM_017005650.1 | XP_016861139.1 | 13 - 14 | 2172 | 93 | |
XM_017005651.1 | XP_016861140.1 | 13 - 14 | 1698 | 93 | |
XM_017005652.1 | XP_016861141.1 | 15 - 16 | 2172 | 93 | |
XM_017005653.1 | XP_016861142.1 | 3 - 4 | 650 | 93 | |
GenBank mRNA | AF109737.1 | - | 12 - 13 | 2041 | 93 |
AF150882.1 | - | 12 - 13 | 2041 | 93 | |
AF188679.1 | - | 13 - 14 | 1872 | 93 | |
AJ417790.1 | - | 12 - 13 | 1842 | 93 |