Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014332.2 | NP_055147.1 | 2 - 3 | 278 | 85 |
NR_045617.1 | 2 - 3 | 278 | 85 | ||
GenBank mRNA | AF129505.1 | - | 2 - 3 | 230 | 85 |
AJ250584.1 | - | 2 - 3 | 230 | 85 | |
AK312134.1 | - | 2 - 3 | 278 | 85 | |
BC005948.1 | - | 2 - 3 | 159 | 85 | |
BF126275.1 | - | 2 - 3 | 140 | 85 |