Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001485.2 | NP_001001485.1 | 2 - 3 | 507 | 81 |
NM_001001486.1 | NP_001001486.1 | 1 - 2 | 184 | 81 | |
NM_001001487.1 | NP_001001487.1 | 1 - 2 | 184 | 81 | |
NM_001199179.1 | NP_001186108.1 | 2 - 3 | 413 | 81 | |
NM_001199185.1 | NP_001186114.1 | 1 - 2 | 326 | 81 | |
NM_014382.3 | NP_055197.2 | 1 - 2 | 326 | 81 | |
XM_005247355.2 | XP_005247412.1 | 2 - 3 | 554 | 81 | |
XM_005247356.2 | XP_005247413.1 | 2 - 3 | 307 | 81 | |
XM_017006164.1 | XP_016861653.1 | 2 - 3 | 215 | 81 | |
GenBank mRNA | AB037768.1 | - | 2 - 3 | 413 | 82 |
AF181120.1 | - | 1 - 2 | 239 | 81 | |
AF181121.1 | - | 1 - 2 | 239 | 81 | |
AK314342.1 | - | 1 - 2 | 188 | 81 | |
AY268374.1 | - | 1 - 2 | 133 | 81 | |
AY268375.1 | - | 1 - 2 | 133 | 81 | |
BC028139.1 | - | 1 - 2 | 196 | 81 | |
DC343471.1 | - | 1 - 2 | 326 | 81 |