Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278722.1 | NP_001265651.1 | 3 - 4 | 541 | 98 |
NM_014432.3 | NP_055247.3 | 3 - 4 | 860 | 98 | |
XM_017010954.1 | XP_016866443.1 | 3 - 4 | 1049 | 98 | |
XM_017010955.1 | XP_016866444.1 | 3 - 4 | 509 | 98 | |
GenBank mRNA | AF184971.1 | - | 3 - 4 | 644 | 98 |
AK297121.1 | - | 3 - 4 | 503 | 98 | |
BC113574.1 | - | 3 - 4 | 738 | 98 | |
BC113602.1 | - | 3 - 4 | 738 | 98 |