Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014574.3 | NP_055389.3 | 7 - 8 | 1207 | 71 |
GenBank mRNA | AK290229.1 | - | 7 - 8 | 1185 | 71 |
AK314041.1 | - | 7 - 8 | 1173 | 71 | |
BC126221.1 | - | 7 - 8 | 1125 | 71 | |
BC132673.1 | - | 7 - 8 | 1125 | 71 | |
U17989.1 | - | 7 - 8 | 1195 | 71 |