Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014654.3 | NP_055469.3 | 3 - 4 | 917 | 61 |
XM_011542463.1 | XP_011540765.1 | 3 - 4 | 852 | 61 | |
XM_011542464.2 | XP_011540766.1 | 3 - 4 | 1122 | 61 | |
XM_011542466.1 | XP_011540768.1 | 3 - 4 | 759 | 61 | |
GenBank mRNA | AB007937.1 | - | 1 - 2 | 2199 | 61 |
AF248634.1 | - | 3 - 4 | 924 | 61 | |
AK295698.1 | - | 2 - 3 | 662 | 61 | |
BQ887307.1 | - | 1 - 2 | 235 | 61 | |
KU178484.1 | - | 2 - 3 | 654 | 61 | |
KU178485.1 | - | 1 - 2 | 696 | 61 |